Skip to Content
Merck
All Photos(1)

Key Documents

578754

Sigma-Aldrich

N-(4-Phenylboronic)succinamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(OH)2BC6H4NHCOCH2CH2CO2H
CAS Number:
Molecular Weight:
237.02
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

165-169 °C (lit.)

functional group

amide
carboxylic acid

SMILES string

OB(O)c1ccc(NC(=O)CCC(O)=O)cc1

InChI

1S/C10H12BNO5/c13-9(5-6-10(14)15)12-8-3-1-7(2-4-8)11(16)17/h1-4,16-17H,5-6H2,(H,12,13)(H,14,15)

InChI key

VYUCUZOJUHAJLU-UHFFFAOYSA-N

Application

Reactant for:
  • Construction of combinatorial artificial receptor array (CARA)
  • Preparation of hydroxycoumarins and pyranocoumarins via Pechmann condensation and Pd-catalyzed Suzuki coupling as antitumor agents

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service