Skip to Content
Merck
All Photos(2)

Documents

572659

Sigma-Aldrich

Germanium(II) bromide

97%

Synonym(s):

Dibromogermylene, Germanium dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GeBr2
CAS Number:
Molecular Weight:
232.45
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: germanium

mp

120-125 °C (lit.)

SMILES string

Br[GeH2]Br

InChI

1S/Br2GeH2/c1-3-2/h3H2

InChI key

JPSUHKQGIXBUKX-UHFFFAOYSA-N

Application

  • Germanium speciation in experimental and natural sphalerite: The research tests the impact of different oxidation states of germanium, including Ge(II) bromide, on the enrichment processes in hydrothermal Zn-Pb ores (Liu et al., 2023).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service