Skip to Content
Merck
All Photos(1)

Key Documents

569828

Sigma-Aldrich

Pyridinium dichromate on silica gel

200-400 mesh, extent of labeling: 20 wt. % silica gel loading

Synonym(s):

PDC on silica

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

extent of labeling

20 wt. % silica gel loading

particle size

200-400 mesh

storage temp.

2-8°C

SMILES string

c1cc[nH+]cc1.c2cc[nH+]cc2.[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

InChI

1S/2C5H5N.2Cr.7O/c2*1-2-4-6-5-3-1;;;;;;;;;/h2*1-5H;;;;;;;;;/q;;;;;;;;;2*-1/p+2

InChI key

LMYWWPCAXXPJFF-UHFFFAOYSA-P

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B Inhalation - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functionalized silica gel is used in size-exclusion chromatography of cationic polymers and in the separation of sugars, steroids, cholesterol, and triglycerides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service