Skip to Content
Merck
All Photos(1)

Documents

566187

Sigma-Aldrich

Indenylzirconium(IV) trichloride

97%

Synonym(s):

Trichloro[(1,2,3,3a,7a-η)-1H-inden-1-yl]zirconium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl3Zr
CAS Number:
Molecular Weight:
312.73
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

reagent type: catalyst
core: zirconium

mp

151-161 °C (lit.)

SMILES string

Cl[Zr](Cl)Cl.[CH]1[CH][C]2C=CC=C[C]2[CH]1

InChI

1S/C9H7.3ClH.Zr/c1-2-5-9-7-3-6-8(9)4-1;;;;/h1-7H;3*1H;/q;;;;+3/p-3

InChI key

RHUATJQAOSHYRG-UHFFFAOYSA-K

Application

Reactant for:
  • Synthesis of zirconocenes bearing benz[f]indenyl ligand
  • Monoindenyl halides of zirconium

Polymerization catalyst for modification of polyethylene polydiversity via blending Ziegler-Natta catalysts with IV-half metallocene or scorpionate complexes

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service