56194
4-Hydroxyphthalonitrile
technical, ≥90% (GC)
Synonym(s):
4-Hydroxybenzene-1,2-dicarbonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H4N2O
CAS Number:
Molecular Weight:
144.13
Beilstein:
2718847
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Assay
≥90% (GC)
form
solid
mp
~218 °C
SMILES string
Oc1ccc(C#N)c(c1)C#N
InChI
1S/C8H4N2O/c9-4-6-1-2-8(11)3-7(6)5-10/h1-3,11H
InChI key
FTVOPKROFUTOKY-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Young-Keun Kim et al.
International journal of molecular sciences, 9(12), 2745-2756 (2009-03-31)
We have synthesized soluble TiOPc derivatives containing alkoxy groups for use as additives in dye-sensitized solar cells (DSSCs). The DSSC devices containing these TiOPc derivatives exhibited short-circuit current densities of 8.49~10.04 mA/cm(2) and power conversion efficiencies of 2.73~3.62 % under
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service