Skip to Content
Merck
All Photos(1)

Key Documents

560448

Sigma-Aldrich

2,3-Dimethoxyphenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2NH2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

drug control

USDEA Schedule I

refractive index

n20/D 1.5320 (lit.)

bp

261-262 °C (lit.)

density

1.178 g/mL

SMILES string

COc1cccc(CCN)c1OC

InChI

1S/C10H15NO2/c1-12-9-5-3-4-8(6-7-11)10(9)13-2/h3-5H,6-7,11H2,1-2H3

InChI key

XKBUFTXNLBWTFP-UHFFFAOYSA-N

General description

2,3-Dimethoxyphenethylamine is identified as one of the components in bamboo (Phyllostachys pubescens) plant extract by gas chromatographic- mass spectrometric analysis.[1] 1-(2,3-dimethoxyphenyl)-2-nitroethene undergoes reduction reaction with lithium aluminum hydride (LiAlH4) in the presence of dry tetrahydrofuran to yield 2,3-dimethoxyphenethylamine.[2]

Application

2,3-Dimethoxyphenethylamine may be used in the synthesis of N-(2,3-dimethoxyphenethyl)-2-(3,4-dimethoxyphenyl)acetamide[2] and N-(3,4-dimethoxyphenethyl)-phenyl acetamide.[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-mass spectrometry analysis and chemical composition of the bamboo-carbonized liquid.
Bilehal D, et al.
Food Analytical Methods, 5(1), 109-112 (2012)
Tetramethoxydibenzoquinolizinium salts. Preparation and antileukemic activity of some positional and structural isomers of coralyne.
Zee-Cheng RKY and Cheng CC.
Journal of Medicinal Chemistry, 19(7), 882-886 (1976)
Chen-Yuan Kuo et al.
European journal of medicinal chemistry, 44(3), 1271-1277 (2008-10-15)
A series of phenoxyisoquinolines, N-phenoxyethyl-1-(2-nitrophenyl)-1,2,3,4-THIQs 3a-3d, N-phenoxyethyl-1-benzyl-1,2,3,4-THIQ 3e, N-phenoxyethyl-1-(2-aminophenyl)-1,2,3,4-THIQs 5f-5i, N-phenoxyethyl-1-(2-phenoxyethylaminophenyl)-1,2,3,4-THIQs 5f'-5i', have been synthesized and tested in isolated rat vas deferens alpha-adrenoreceptors. Comparison of pA2 values for these compounds in the presence of phenylephrine confirms that alpha(1)-adrenoceptor blocking activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service