543233
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt bound to PEG 750
Synonym(s):
Burgess reagent on PEG, PEG supported Burgess reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: proton reactive
SMILES string
OCCO.CC[N+](CC)(CC)S(=O)(=O)[N-]C(=O)OCCO
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service