Skip to Content
Merck
All Photos(1)

Key Documents

543055

Sigma-Aldrich

Ethyl 1-cyano-1-cyclopropanecarboxylate

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€35.42

€35.42

List Price€50.60Save 30%

In StockDetails


Request a Bulk Order

Select a Size

Change View
5 G
€35.42

About This Item

Linear Formula:
NCC3H4CO2C2H5
CAS Number:
Molecular Weight:
139.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€35.42

List Price€50.60Save 30%

In StockDetails


Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.445 (lit.)

bp

217 °C (lit.)

density

1.077 g/mL at 25 °C (lit.)

functional group

ester
nitrile

SMILES string

CCOC(=O)C1(CC1)C#N

InChI

1S/C7H9NO2/c1-2-10-6(9)7(5-8)3-4-7/h2-4H2,1H3

InChI key

FDZLCIITHLSEQK-UHFFFAOYSA-N

General description

Ethyl 1-cyano-1-cyclopropanecarboxylate (ethyl 1-cyanocyclopropanecarboxylate) is an alkyl 1-cyanocyclopanecarboxylate. It can be prepared by reacting ethyl cyanoacetate, 1,2-dibromoethane, potassium carbonate and DMSO.[1]

Application

Ethyl 1-cyano-1-cyclopropanecarboxylate may be used as a monomer in the preparation of poly(ethyl trimethylene-1-cyano-1-carboxylate)s.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anionic ring-opening polymerization of alkyl 1-cyanocyclopropanecarboxylates
Kagumba LC and Penelle J.
Macromolecules, 38.11, 4588-4594 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service