Skip to Content
Merck
All Photos(1)

Key Documents

542393

Sigma-Aldrich

4-Methyl-3-thienylboronic acid

≥95%

Synonym(s):

(4-Methylthien-3-yl)boronic acid, (4-Methylthiophen-3-yl)boronic acid, (4-Methylthiophen-3-yl)dihydroxyborane, 4-Methyl-3-thiopheneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7BO2S
CAS Number:
Molecular Weight:
141.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

mp

138-144 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1cscc1B(O)O

InChI

1S/C5H7BO2S/c1-4-2-9-3-5(4)6(7)8/h2-3,7-8H,1H3

InChI key

LVPFZXKLROORIK-UHFFFAOYSA-N

Application

Reactant for preparation of biologically active molecules:
  • Selective sphingosine phosphate receptor antagonists
  • Biarylimidazole derivatives as H+/K+ ATPase inhibitors
  • Benzothienopyrimidinones as inhibitors of human protooncogene proviral insertion site in moloney murine leukemia virus kinases

Used in:
  • Synthesis of arene-functionalized fullerenes via rhodium-catalyzed hydroarylation
  • Suzuki cross-coupling reactions

Analysis Note

May contain up to 1 equivalent of water

Other Notes

Contains varying amounts of anhydride.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service