Skip to Content
Merck
All Photos(1)

Key Documents

541761

Sigma-Aldrich

Copper bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)

≥99.99% trace metals basis

Synonym(s):

Copper(II) (6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate), Cu(FOD)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(OCC(CH3)3CHCOCF2CF2CF3)2
CAS Number:
Molecular Weight:
653.89
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.99% trace metals basis

form

solid

reaction suitability

core: copper

mp

59-61 °C (lit.)

SMILES string

CC(C)(C)C(=O)\C=C(/O[Cu]O\C(=C/C(=O)C(C)(C)C)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/2C10H11F7O2.Cu/c2*1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h2*4,19H,1-3H3;/q;;+2/p-2/b2*6-4-;

InChI key

OXBBKSQDOOZZFZ-IHJILFKMSA-L

General description

Atomic number of base material: 29 Copper

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service