Recommended Products
Assay
97%
refractive index
n20/D 1.456 (lit.)
bp
111-113 °C/0.2 mmHg (lit.)
density
1.234 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)CS(C)(=O)=O
InChI
1S/C5H10O4S/c1-3-9-5(6)4-10(2,7)8/h3-4H2,1-2H3
InChI key
OCCWQCYBCZADCE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Ethyl methylsulfonylacetate is an active methylene compound.
Application
Ethyl methylsulfonylacetate may be used in the synthesis of:
- 3-methylamino-5-phenyl-2-thiophenecarboxylate
- 3-(methylsulfonyl)-2H-chromen-2-one
- 7-hydroxy-3-(methylsulfonyl)-2H-chromen-2-one
- 7-bromo-3-(methylsulfonyl)-2H-chromen-2-one
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
228.2 °F - closed cup
Flash Point(C)
109 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rhodium (II)-mediated reactions of thiobenzoylketene S, N-acetals with a-diazo carbonyl compounds: synthesis of 2-substituted 3-alkylamino-5-phenylthiophenes.
Journal of the Chemical Society. Perkin Transactions 1, 21, 2414-2417 (2002)
Yonemitsu-type condensations catalysed by proline and Eu (OTf) 3.
Royal Society of Chemistry Advances, 4(89), 47992-47999 (2014)
Molecules (Basel, Switzerland), 16(6), 4379-4388 (2011-05-31)
The ionic liquid 1,3-dimethylimidazolium methyl sulfate, [MMIm][MSO₄], together with a small amount of water (the amount taken up by the ionic liquid upon exposure to air), acts efficiently as both solvent and catalyst of the Knoevenagel condensation reactions of malononitrile
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service