Skip to Content
Merck
All Photos(1)

Documents

539090

Sigma-Aldrich

2-Chloro-6-fluorobenzyl bromide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CH2Br
CAS Number:
Molecular Weight:
223.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.568 (lit.)

density

1.629 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(Cl)c1CBr

InChI

1S/C7H5BrClF/c8-4-5-6(9)2-1-3-7(5)10/h1-3H,4H2

InChI key

IGUVNNXFTDCASP-UHFFFAOYSA-N

Application

2-Chloro-6-fluorobenzyl bromide [2-(bromomethyl)-1-chloro-3-fluorobenzene] may be used in the synthesis of 4-chlorobenzoimidazo[1,2-a][3,1]benzothiazine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A New Copper-Catalyzed Pathway to Benzo and Pyridyl Fused Imidazo-, Triazolo-and Pyrimido-Thiazines.
Abele E, et al.
Heterocyclic Letters, 2(2), 182-191 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service