All Photos(1)
About This Item
Linear Formula:
ClC6H2(OCH3)2NH2
CAS Number:
Molecular Weight:
187.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
97%
mp
118-120 °C (lit.)
SMILES string
COc1cc(Cl)c(OC)cc1N
InChI
1S/C8H10ClNO2/c1-11-7-4-6(10)8(12-2)3-5(7)9/h3-4H,10H2,1-2H3
InChI key
YGUFQYGSBVXPMC-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - STOT RE 2 Oral
Target Organs
Other,Blood
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
325.4 °F
Flash Point(C)
163 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service