Skip to Content
Merck
All Photos(1)

Key Documents

536202

Sigma-Aldrich

1-Pentyl isocyanide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4NC
CAS Number:
Molecular Weight:
97.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4064 (lit.)

density

0.772 g/mL at 25 °C (lit.)

functional group

amine
isonitrile

storage temp.

2-8°C

SMILES string

CCCCC[N+]#[C-]

InChI

1S/C6H11N/c1-3-4-5-6-7-2/h3-6H2,1H3

InChI key

QVUBMHTYZLUXSP-UHFFFAOYSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

25.0 °F - closed cup

Flash Point(C)

-3.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas C Anderson et al.
Journal of the American Chemical Society, 140(23), 7199-7205 (2018-05-11)
Using 31P nuclear magnetic resonance (NMR) spectroscopy, we monitor the competition between tri- n-butylphosphine (Bu3P) and various amine and phosphine ligands for the surface of chloride terminated CdSe nanocrystals. Distinct 31P NMR signals for free and bound phosphine ligands allow

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service