Skip to Content
Merck
All Photos(1)

Key Documents

532622

Sigma-Aldrich

3-Nitro-2,6-lutidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2
CAS Number:
Molecular Weight:
152.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

26-31 °C (lit.)

SMILES string

Cc1ccc(c(C)n1)[N+]([O-])=O

InChI

1S/C7H8N2O2/c1-5-3-4-7(9(10)11)6(2)8-5/h3-4H,1-2H3

InChI key

AETHUDGJSSKZKT-UHFFFAOYSA-N

General description

3-Nitro-2,6-lutidine, a nitropyridine derivative, is also known as 2,6-dimethyl-3-nitropyridine. It can be prepared from 2,6-lutidine via nitration.

Application

3-Nitro-2,6-lutidine may be used in the preparation of 3-methoxy-2,7-dimethyl-2H-1,4-diazepine1 and 3-amino-2,6-lutidine.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

100.0 °F - closed cup

Flash Point(C)

37.78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Methylpyridine Derivatives (XXI). Amination of Chloro-2,6-lutidines and Their N-Oxides via a Hetaryne Mechanism.
Kato T and Niitsuma T.
Chemical & Pharmaceutical Bulletin, 13(8), 963-968 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service