Skip to Content
Merck
All Photos(2)

Key Documents

528099

Sigma-Aldrich

8-Amino-6-methoxyquinoline hydrobromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O · HBr
CAS Number:
Molecular Weight:
255.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

238 °C (dec.) (lit.)

SMILES string

Br.COc1cc(N)c2ncccc2c1

InChI

1S/C10H10N2O.BrH/c1-13-8-5-7-3-2-4-12-10(7)9(11)6-8;/h2-6H,11H2,1H3;1H

InChI key

WGYQQCWLZRGIKB-UHFFFAOYSA-N

General description

8-Amino-6-methoxyquinoline hydrobromide is a quinoline building block.[1] It is formed during the synthesis of α-morpholino-β-(N-8-amino-6-methoxyquinoline)-benzylacetophenone.[2]

Application

8-Amino-6-methoxyquinoline hydrobromide may be used as an amino component in a novel series of bladder-selective diaminocyclobutenedione potassium channel openers.[3] It may also be used for the synthesis of 1-N-(6-methoxy-8-quinoyl)-4′-carboxyl-benzensulfonamide.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

a, ?-Diamino Ketones. III. 1 Reaction of 8-Amino-6-methoxyquinoline with a-Bromo-?-aminoketones.
Cromwell NH and Hoeksema H.
Journal of the American Chemical Society, 67(1), 124-125 (1945)
Butera JA.
Journal of Medicinal Chemistry, 43, 1187-1187 (2000)
Armer RE, et al.
Bioorganic & Medicinal Chemistry, 9, 2430-2430 (1999)
Iris Boll et al.
Bioorganic & medicinal chemistry letters, 16(10), 2781-2785 (2006-02-24)
Two ligand-intercalator-peptide nucleic acid conjugates (L-NADI-PNAs) have been synthesized. Affinity of these conjugates to their complementary DNAs was found to be affected by Zn(2+). The magnitude of this effect could be controlled by a variation of the ligand. Upon binding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service