Skip to Content
Merck
All Photos(1)

Key Documents

526061

Sigma-Aldrich

Diethyl 4-chlorobenzylphosphonate

97%

Synonym(s):

p-(Chlorobenzyl)phosphonic acid diethyl ester, NSC 202843

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
262.67
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.509 (lit.)

bp

162 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(Cc1ccc(Cl)cc1)OCC

InChI

1S/C11H16ClO3P/c1-3-14-16(13,15-4-2)9-10-5-7-11(12)8-6-10/h5-8H,3-4,9H2,1-2H3

InChI key

BVQPVBZRJSFOEZ-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of chromenone and chromene derivatives as anti-picornavirus capsid-binders
  • Preparation of wide band gap monomers and silafluorene copolymers with blue electroluminescence, as emitter layer in organic LEDs
  • Asymmetric synthesis of gamma-nitrophosphonates via Michael addition with nitroalkenes
  • Wittig-Horner reactions

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service