Skip to Content
Merck
All Photos(1)

Key Documents

522945

Sigma-Aldrich

2′,6′-Dichloro-3′-fluoroacetophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(F)COCH3
CAS Number:
Molecular Weight:
207.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.529 (lit.)

bp

255 °C (lit.)

density

1.403 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1c(Cl)ccc(F)c1Cl

InChI

1S/C8H5Cl2FO/c1-4(12)7-5(9)2-3-6(11)8(7)10/h2-3H,1H3

InChI key

VJBFZHHRVCPAPZ-UHFFFAOYSA-N

General description

2′,6′-Dichloro-3′-fluoroacetophenone is an aryl fluorinated building block.

Application

2′,6′-Dichloro-3′-fluoroacetophenone (2,6-Dichloro-3-fluoroacetophenone) may be used to synthesize the enantiomerically pure form of (S)-1-(2,6-dichloro-3-fluorophenyl)ethanol.[1]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Whittall J and Sutton PW.
Practical Methods for Biocatalysis and Biotransformations (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service