Skip to Content
Merck
All Photos(1)

Documents

521620

Sigma-Aldrich

2-Amino-3,5-dinitrothiophene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3N3O4S
CAS Number:
Molecular Weight:
189.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

174-176 °C (lit.)

SMILES string

Nc1sc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C4H3N3O4S/c5-4-2(6(8)9)1-3(12-4)7(10)11/h1H,5H2

InChI key

DZRZHFOFVWAKGT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of highly active thiophene ring-containing chromophore components for photonic polymers based on a newly designed route.
Yuquan S, et al.
Journal of the Chemical Society. Perkin Transactions 1, 24, 3691-3695 (1999)
Synthesis and characterization of hetarylazo disperse dyes derived from substituted N-?-acetoxyethylanilines-Application on cellulose acetate.
Georgiadou KL, et al.
Journal of Applied Polymer Science, 92(6), 3479-3483 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service