All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
bp
105-107 °C/6 mmHg (lit.)
mp
41-43 °C (lit.)
SMILES string
[O-][N+](=O)c1cccc2CCCc12
InChI
1S/C9H9NO2/c11-10(12)9-6-2-4-7-3-1-5-8(7)9/h2,4,6H,1,3,5H2
InChI key
ZQABKMBXKCJTPM-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 235-235 (2015)
Details of the steric interaction in mono-ortho-substituted nitrobenzene anion radical.
McKinney TM and Geske DH.
Journal of the American Chemical Society, 89(12), 2806-2813 (1967)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service