Skip to Content
Merck
All Photos(1)

Key Documents

513911

Sigma-Aldrich

4-Methylbenzenethiol sodium salt

98%

Synonym(s):

4-Thiocresol sodium salt, Sodium 4-methylphenylthiolate, p-Methylthiophenol sodium salt, p-Thiocresol sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€184.00

€184.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
€184.00

About This Item

Linear Formula:
CH3C6H4SNa
CAS Number:
Molecular Weight:
146.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€184.00


Available to ship on16 April 2025Details


Request a Bulk Order

Assay

98%

mp

>300 °C (lit.)

SMILES string

Cc1ccc(S[Na])cc1

InChI

1S/C7H8S.Na/c1-6-2-4-7(8)5-3-6;/h2-5,8H,1H3;/q;+1/p-1

InChI key

PRORQUQEDGGDQO-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metallation reactions. Part 35: A change of the regiochemistry in the metallation of (alkylthio) arenes.
Cabiddu MG, et al.
Tetrahedron, 3915-3920 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service