Skip to Content
Merck
All Photos(1)

Key Documents

495964

Sigma-Aldrich

3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one

98%

Synonym(s):

DEPBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N3O5P
CAS Number:
Molecular Weight:
299.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

72-75 °C (lit.)

SMILES string

CCOP(=O)(OCC)ON1N=Nc2ccccc2C1=O

InChI

1S/C11H14N3O5P/c1-3-17-20(16,18-4-2)19-14-11(15)9-7-5-6-8-10(9)12-13-14/h5-8H,3-4H2,1-2H3

InChI key

AJDPNPAGZMZOMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one (DEPBT) is a coupling reagent that facilitates amide bond formation and also resists racemization. It has been shown to be more effective than phosphonium and uronium coupling reagents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H Li et al.
Organic letters, 1(1), 91-93 (2000-05-24)
[formula: see text] The new crystalline phosphate reagent 3-(diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one (DEPBT) mediates amide bond formation with a remarkable resistance to racemization. Comparative racemization studies were carried out and DEPBT proved to be superior to typical phosphonium and uronium coupling reagents. DEPBT
Yun-Hua Ye et al.
Biopolymers, 80(2-3), 172-178 (2005-01-01)
3-(Diethoxyphosphoryloxy)-1, 2, 3-benzotriazin-4(3H)-one (DEPBT) is an effective coupling reagent for synthesis of linear and cyclic peptides by both solution and solid-phase peptide synthesis. DEPBT mediates amide bond formation with remarkable resistance to racemization. When DEPBT is used as a coupling
New reagents, reactions, and peptidomimetics for drug design.
Goodman M, et al.
Pept. Sci., 60(3), 229-245 (2001)
Daniel Herkommer et al.
ChemMedChem, 10(3), 470-489 (2015-02-03)
The natural products rhizopodin and bistramide belong to an elite class of highly potent actin binding agents. They show powerful antiproliferative activities against a range of tumor cell lines, with IC50 values in the low-nanomolar range. At the molecular level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service