Skip to Content
Merck
All Photos(1)

Key Documents

479888

Sigma-Aldrich

3,4,5-Trimethoxybenzaldehyde dimethyl acetal

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CH(OCH3)2
CAS Number:
Molecular Weight:
242.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

98%

refractive index

n20/D 1.514 (lit.)

bp

180 °C/10 mmHg (lit.)

density

1.147 g/mL at 25 °C (lit.)

SMILES string

COC(OC)c1cc(OC)c(OC)c(OC)c1

InChI

1S/C12H18O5/c1-13-9-6-8(12(16-4)17-5)7-10(14-2)11(9)15-3/h6-7,12H,1-5H3

InChI key

PEZMUQHWXSFOIK-UHFFFAOYSA-N

General description

3,4,5-Trimethoxybenzaldehyde dimethyl acetal can be prepared by reacting 3,4,5-trimethoxybenzaldehyde with trimethyl orthoformate.[1]

Application

3,4,5-Trimethoxybenzaldehyde dimethyl acetal may be used to synthesize 3,5-dimethoxy-4-ethylbenzaldehyde, which is an intermediate for preparing 4-alkyl-3,5-dihydroxy-substituted trans-stilbenes.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of antibiotic stilbenes by reductive metalation of 3,4,5-trimethoxybenzaldehyde dimethyl acetal.
Azzena U, et al.
Synthetic Communications, 33(8), 1309-1317 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service