Skip to Content
Merck
All Photos(1)

Key Documents

472778

Sigma-Aldrich

3,4-Di-O-acetyl-6-O-(tert-butyldimethylsilyl)-D-glucal

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O6Si
CAS Number:
Molecular Weight:
344.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

Assay

96%

optical activity

[α]20/D −7°, c = 1.4 in chloroform

refractive index

n20/D 1.456 (lit.)

density

1.044 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)O[C@H]([C@H](O)CO[Si](C)(C)C(C)(C)C)[C@H](OC(C)=O)[C@@H](O)C=O

InChI

1S/C16H30O8Si/c1-10(18)23-14(12(20)8-17)15(24-11(2)19)13(21)9-22-25(6,7)16(3,4)5/h8,12-15,20-21H,9H2,1-7H3/t12-,13+,14+,15+/m0/s1

InChI key

DGPMSALVNCCUAW-GBJTYRQASA-N

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service