Skip to Content
Merck

469459

Sigma-Aldrich

Boc-D-valinol

98%

Synonym(s):

N-(tert-Butoxycarbonyl)-D-valinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
203.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Assay

98%

optical activity

[α]23/D +23°, c = 1 in chloroform

reaction suitability

reaction type: Boc solid-phase peptide synthesis

refractive index

n20/D 1.451 (lit.)

bp

218 °C

density

0.995 g/mL at 25 °C

application(s)

peptide synthesis

SMILES string

CC(C)[C@H](CO)NC(=O)OC(C)(C)C

InChI

1S/C10H21NO3/c1-7(2)8(6-12)11-9(13)14-10(3,4)5/h7-8,12H,6H2,1-5H3,(H,11,13)/t8-/m0/s1

InChI key

OOQRRYDVICNJGC-QMMMGPOBSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
AV43517MABS508HPA011008
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

UniProt accession no.

P00505

UniProt accession no.

P00505

UniProt accession no.

P17174

UniProt accession no.

Q14789

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service