Skip to Content
Merck
All Photos(1)

Key Documents

464147

Sigma-Aldrich

3-Iodo-4,5-dimethoxybenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H2(OCH3)2CHO
CAS Number:
Molecular Weight:
292.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

69-72 °C (lit.)

functional group

aldehyde
iodo

SMILES string

COc1cc(C=O)cc(I)c1OC

InChI

1S/C9H9IO3/c1-12-8-4-6(5-11)3-7(10)9(8)13-2/h3-5H,1-2H3

InChI key

MVPNBXPAUYYZAF-UHFFFAOYSA-N

General description

3-Iodo-4,5-dimethoxybenzaldehyde is an aryl iodide having an aldehyde group. It can be synthesized starting from 3,4-dihydroxy-5-iodobenzaldehyde.[1]

Application

3-Iodo-4,5-dimethoxybenzaldehyde was used in the preparation of 3-iodo-4,4′,5-trimethoxy-3′-O-tert-butyldiphenylsilyl-Z-stilbene and 3-iodo-4,4′,5-trimethoxy-3′-O-tert-butyldiphenylsilyl-E-stilbene.[2] It may be used for the synthesis of chloropeptin I and chloropeptin II.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George R Pettit et al.
Journal of natural products, 75(3), 385-393 (2012-02-14)
Toward the objective of designing a structurally modified analogue of the combretastatin A-4 phosphate prodrug (1b) with the potential for increased specificity toward thyroid carcinoma, synthesis of a series of iodocombstatin phosphate (11a-h) and diiodocombstatin phosphate prodrugs (12a-h) has been
Joie Garfunkle et al.
Journal of the American Chemical Society, 131(44), 16036-16038 (2009-10-21)
The first total synthesis of chloropeptin II (1, complestatin) is disclosed. Key elements of the approach include the use of an intramolecular Larock indole synthesis for the initial macrocyclization, adopting conditions that permit utilization of a 2-bromoaniline, incorporating a terminal
Approaches to iodinated derivatives of vanillin and isovanillin.
Nammalwar B, et al.
Organic preparations and procedures international, 44(2), 146-146 (2012)
Nucleophilic catalysis of the iodine-zinc exchange reaction: preparation of highly functionalized diaryl zinc compounds.
Florian F Kneisel et al.
Angewandte Chemie (International ed. in English), 43(8), 1017-1021 (2004-02-18)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service