Skip to Content
Merck
All Photos(1)

Key Documents

462780

Sigma-Aldrich

Barium tetracyanoplatinate(II) hydrate

99.9%

Synonym(s):

Tetrakis(cyano-κC)platinate barium hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaPt(CN)4 · xH2O
CAS Number:
Molecular Weight:
436.47 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9%

reaction suitability

core: barium
reagent type: catalyst

mp

100 °C (dec.) (lit.)

density

3.05 g/mL at 25 °C (lit.)

SMILES string

O.[Ba++].N#C[Pt--](C#N)(C#N)C#N

InChI

1S/4CN.Ba.H2O.Pt/c4*1-2;;;/h;;;;;1H2;/q;;;;+2;;-2

InChI key

BBYLKQFCIDTBFK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service