Skip to Content
Merck
All Photos(1)

Key Documents

462241

Sigma-Aldrich

Potassium pentamethylcyclopentadienide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15K
CAS Number:
Molecular Weight:
174.32
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

mp

>230 °C (lit.)

SMILES string

[K+].Cc1c(C)c(C)[c-](C)c1C

InChI

1S/C10H15.K/c1-6-7(2)9(4)10(5)8(6)3;/h1-5H3;

InChI key

XJLFQRRHDGCZLZ-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Starting materials for low and high-valent uranium chemistry
  • Heterometallic ytterbium-alkali-metal boratabenzene complexes
  • Sandwich complexes of di- and trivalent ytterbium
  • Samarium(II) borohydrides
  • Half-sandwich scandium complexes
  • Rare-earth metal complexes

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service