Skip to Content
Merck
All Photos(1)

Key Documents

462195

Sigma-Aldrich

1,3-Dibutyl-2-thiobarbituric acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N2O2S
CAS Number:
Molecular Weight:
256.36
MDL number:
UNSPSC Code:
12352100

Assay

95%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

61-63 °C (lit.)

SMILES string

CCCCN1C(=O)CC(=O)N(CCCC)C1=S

General description

1,3-Dibutyl-2-thiobarbituric acid (DBTB) is a non-polar barbituric acid derivative.[1] Its electronic structure has been studied by HeI and HeII UV photoelectron spectroscopy (UPS) and quantum chemical calculations.[2]

Application

1,3-Dibutyl-2-thiobarbituric acid has been employed as a derivatization reagent for the determination of nicotine and its metabolites in urine by a quantitative method based on König reaction. This method helps to differentiate between smokers and non-smokers.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electronic structure and biological activity: Barbiturates vs. thiobarbiturates.
Novak I and Kovac B.
Chemical Physics Letters, 493(4), 242-244 (2010)
K L Chambers et al.
British journal of biomedical science, 58(2), 61-65 (2001-07-07)
A simple, quick and inexpensive test for smoking status would be useful in a variety of settings. The non-polar barbituric acid derivative 1,3-dibutyl-2-thiobarbituric acid (DBTB) is described as a novel derivatisation reagent for nicotine and its metabolites in the König

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service