Skip to Content
Merck
All Photos(1)

Documents

458392

Sigma-Aldrich

Poly[di(ethylene glycol) adipate]

average Mn ~2,500

Synonym(s):

Adipic acid-diethylene glycol polyester, Diethylene glycol-adipic acid copolymer, Oligodiethylene glycol adipate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OCH2CH2OCO(CH2)4CO2CH2CH2]nOH
CAS Number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~2,500

refractive index

n20/D 1.476

viscosity

11,000 cP(25 °C)(lit.)

mp

<−14 °C

hydroxyl functionality

2.0

hydroxyl value

45 mg KOH/g

density

1.192 g/mL at 25 °C

SMILES string

OCCO.OC(=O)CCCCC(O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service