Skip to Content
Merck
All Photos(1)

Key Documents

457191

Sigma-Aldrich

2,2′-Bipyridine-3,3′−dicarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2O4
CAS Number:
Molecular Weight:
244.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

functional group

carboxylic acid

SMILES string

OC(=O)c1cccnc1-c2ncccc2C(O)=O

InChI

1S/C12H8N2O4/c15-11(16)7-3-1-5-13-9(7)10-8(12(17)18)4-2-6-14-10/h1-6H,(H,15,16)(H,17,18)

InChI key

KNVZVRWMLMPTTJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of (2, 2'-bipyridine-3, 3'-dicarboxylic acid) dichlorocopper (II) monohydrate.
Goddard R, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 46(1), 33-35 (1990)
First examples of ternary lanthanide 2, 2'-bipyridine-3, 3'-dicarboxylate coordination polymers with zigzag chains structures assembled from lanthanide ions, 2, 2'-bipyridine-3, 3'-dicarboxylate and 1, 10-phenanthroline.
Hu M, et al.
Inorgorganica Chimica Acta, 360(5), 1684-1690 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service