Skip to Content
Merck
All Photos(2)

Key Documents

444707

Sigma-Aldrich

Diethyl 1,3,5-benzenetricarboxylate

95%

Synonym(s):

Diethyl 5-carboxyisophthalate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€115.00

€115.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
€115.00

About This Item

Linear Formula:
HO2CC6H3(CO2C2H5)2
CAS Number:
Molecular Weight:
266.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€115.00


Available to ship on31 March 2025Details


Request a Bulk Order

Assay

95%

form

solid

mp

152-154 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

CCOC(=O)c1cc(cc(c1)C(=O)OCC)C(O)=O

InChI

1S/C13H14O6/c1-3-18-12(16)9-5-8(11(14)15)6-10(7-9)13(17)19-4-2/h5-7H,3-4H2,1-2H3,(H,14,15)

InChI key

GAVXMTRMXPWCCV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service