Skip to Content
Merck
All Photos(1)

Key Documents

441341

Sigma-Aldrich

N,N-Dimethylformamide-d7

99.5 atom % D

Synonym(s):

DMF-d7, Heptadeutero-N,N-dimethylformamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCON(CD3)2
CAS Number:
Molecular Weight:
80.14
Beilstein:
1908468
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

99.5 atom % D

Quality Level

Assay

99% (CP)

form

liquid

packaging

pk of 10 × 0.75 mL

technique(s)

NMR: suitable

refractive index

n20/D 1.428 (lit.)

bp

153 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

mass shift

M+7

SMILES string

[2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3,3D

InChI key

ZMXDDKWLCZADIW-YYWVXINBSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung Bin Baek et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(46), 14156-14161 (2015-11-19)
Crystallographic observation of adsorbed gas molecules is a highly difficult task due to their rapid motion. Here, we report the in situ single-crystal and synchrotron powder X-ray observations of reversible CO2 sorption processes in an apparently nonporous organic crystal under
Małgorzata Fabijańska et al.
Dalton transactions (Cambridge, England : 2003), 44(3), 938-947 (2014-08-12)
This paper describes the synthesis of trans-bis-(3-aminoflavone)dichloridoplatinum(ii) (trans-Pt(3-af)2Cl2; TCAP) for use as a potential anticancer compound, and the evaluation of its structure by elemental and spectral analyses, and X-ray crystallography. The complex demonstrated a significant cytotoxic effect against human and
Sean E Doris et al.
Journal of the American Chemical Society, 136(44), 15702-15710 (2014-10-11)
Cationic naked nanocrystals (NCs) are useful building units for assembling hierarchical mesostructured materials. Until now, their preparation required strongly electrophilic reagents that irreversibly sever bonds between native organic ligands and the NC surface. Colloidal instabilities can occur during ligand stripping

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service