Skip to Content
Merck
All Photos(1)

Key Documents

440450

Sigma-Aldrich

3,5-Dinitrophenyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3NCO
CAS Number:
Molecular Weight:
209.12
Beilstein:
3097358
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

82-87 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cc(cc(c1)[N+]([O-])=O)N=C=O

InChI

1S/C7H3N3O5/c11-4-8-5-1-6(9(12)13)3-7(2-5)10(14)15/h1-3H

InChI key

JZPRXQSCMBDGKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthracene derivatives bearing two urea groups as fluorescent receptors for anions.
Kim SK, et al.
Tetrahedron, 61(19), 4545-4550 (2005)
High-performance liquid chromatographic separation of chiral alcohols on chiral stationary phases.
Oi N and Kitahara H.
Journal of Chromatography A, 265, 117-120 (1983)
A J Bourque et al.
Journal of pharmaceutical and biomedical analysis, 11(6), 495-503 (1993-06-01)
Solid-phase derivatization reagents containing a 3,5-dinitrophenyl moiety for the derivatization of amines are described. The reagents are useful for Pirkle-type recognition of stereochemical composition of amines and related nucleophiles. The ability to stabilize 3,5-dinitrophenylisocyanate by covalent immobilization on a polymeric
J N Zeng et al.
Journal of chromatography. B, Biomedical applications, 654(2), 231-248 (1994-04-01)
A new chiral high-performance liquid chromatographic (HPLC) method utilizing ultraviolet (UV) detection has been developed for determining plasma and urinary concentrations of d-fenfluramine and its major metabolite d-norfenfluramine, while being able to determine the possible presence of l-fenfluramine after oral
Sadegh Yaghoubnejad et al.
Journal of separation science, 41(9), 1903-1912 (2018-01-16)
We report the synthesis and enantioseparation characteristics of two novel covalently immobilized deoxycholic acid derivatives as chiral stationary phases for high-performance liquid chromatography. In the structure of the first stationary phase, the 3-position of deoxycholic acid is substituted with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service