439290
2,6-Dichloro-1-fluoropyridinium tetrafluoroborate
97%
Synonym(s):
1-Fluoro-2,6-dichloropyridinium tetrafluoroborate, 2,6-Dichloro-1-fluoropyridin-1-ium, 2,6-Dichloro-1-fluoropyridinium tetrafluoroborate(1-), N-Fluoro-2,6-dichloropyridinium tetrafluoroborate, tetrafluoroboranuide
About This Item
Recommended Products
Quality Level
Assay
97%
mp
212-215 °C (lit.)
functional group
chloro
SMILES string
F[B-](F)(F)F.F[n+]1c(Cl)cccc1Cl
InChI
1S/C5H3Cl2FN.BF4/c6-4-2-1-3-5(7)9(4)8;2-1(3,4)5/h1-3H;/q+1;-1
InChI key
JVYBYXWHPIEUOT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service