Skip to Content
Merck
All Photos(1)

Key Documents

437719

Sigma-Aldrich

Pyridine-d5

99 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Pentadeuteropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5N
CAS Number:
Molecular Weight:
84.13
Beilstein:
114377
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99 atom % D

Assay

99% (CP)

contains

0.03 % (v/v) TMS

refractive index

n20/D 1.506 (lit.)

bp

114.4 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1nc([2H])c([2H])c([2H])c1[2H]

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D

InChI key

JUJWROOIHBZHMG-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Pang et al.
Steroids, 93, 68-76 (2014-12-03)
For the first time, a systematic phytochemical study was performed on the roots of Marsdenia tenacissima. Finally, sixteen new polyoxypregnane glycosides, marstenacissides A1-A7 (1-7) and marstenacissides B1-B9 (8-16), were isolated from M. tenacissima roots. The structures of these new compounds
Xu Pang et al.
Carbohydrate research, 402, 236-240 (2014-12-17)
Five new steroidal saponins (1-5) were isolated from the fermentation broth of total furostanol glycosides from tubers of Dioscorea zingiberensis C.H. Wright incubated with a fungal, Absidia coerulea AS 3.3389, along with known saponins, zingiberensis new saponin (6), deltonin (7)
Indra Prakash et al.
Natural product communications, 9(8), 1135-1138 (2014-09-23)
We report the isolation and complete structure of an isomer of rebaudioside D, known as rebaudioside D2. This novel steviol glycoside was isolated from a bioconversion reaction of rebaudioside A to rebaudioside D. Rebaudioside D2 possesses a relatively rare 1
Shao-Dan Chen et al.
Fitoterapia, 102, 67-73 (2015-02-24)
Five new diarylheptanoids (1-5), along with nine known ones (6-14), were isolated from the rhizomes of Curcuma kwangsiensis. Their structures were established on the basis of spectroscopic analyses. Compounds 1-3 were cyclic diarylheptanoids rarely discovered from C. kwangsiensis. Of all
Yu Shan et al.
Molecules (Basel, Switzerland), 20(11), 20334-20340 (2015-11-17)
A new nortriterpene saponin, 3-O-β-d-glucuronopyranosyl-30-norolean-12,20(29)-dien-23- oxo-28-oic acid, namely bigelovii D (11), was isolated from the hydroalcoholic extract of herbs of Salicornia bigelovii along with 10 known saponins (1-10). Their chemical structures were identified on the basis of spectroscopic analyses including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service