Skip to Content
Merck
All Photos(1)

Key Documents

437069

Supelco

Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide

≥98%, ChiraSelect

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H2(NO2)2NH[CHCH(CH3)2]CONH2
CAS Number:
Molecular Weight:
300.24
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

product line

ChiraSelect

Assay

≥98%

optical activity

[α]20/D +14.0°, c = 2 in acetone

optical purity

ee: ≥99

mp

173-177 °C (lit.)

SMILES string

CC(C)[C@H](Nc1cc(F)c(cc1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C11H13FN4O5/c1-5(2)10(11(13)17)14-7-3-6(12)8(15(18)19)4-9(7)16(20)21/h3-5,10,14H,1-2H3,(H2,13,17)/t10-/m0/s1

InChI key

ZTFFRKNPAXXEBL-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful for the analysis of chiral amino acids by HPLC. Valine analog of Marfey′s reagent that provides improved resolution in the separation of D- and L-amino acids as well as analogs of phenylalanine and proline. It has also aided in understanding the separation mechanism of these reagents.

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harada, K. et al.
Tetrahedron Letters, 36, 1515-1515 (1995)
Chem. Pept. Proteins Part A, 5/6, 145-145 (1993)
Brueckner, H. Keller-Hoehl, Ch. Wittner, R.
Chem. Pept. Proteins Part A, 5/6, 145-145 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service