Skip to Content
Merck
All Photos(1)

Key Documents

434426

Sigma-Aldrich

Ethylbenzene-d10

98 atom % D

Synonym(s):

Decadeuteroethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5CD2CD3
CAS Number:
Molecular Weight:
116.23
Beilstein:
1960387
EC Number:
MDL number:
UNSPSC Code:
12142201

isotopic purity

98 atom % D

refractive index

n20/D 1.492 (lit.)

bp

134.6 °C (lit.)

mp

-95 °C (lit.)

density

0.949 g/mL at 25 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3/i1D3,2D2,3D,4D,5D,6D,7D

InChI key

YNQLUTRBYVCPMQ-CFTAVCBPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in prescored ampules,

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service