Skip to Content
Merck
All Photos(2)

Key Documents

434051

Sigma-Aldrich

Terbium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate)

98%

Synonym(s):

Tb(TMHD)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tb(OCC(CH3)3CHCOC(CH3)3)3
CAS Number:
Molecular Weight:
708.73
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

reaction suitability

core: terbium
reagent type: catalyst

mp

163-164 °C (lit.)

SMILES string

CC(C)(C)C(=O)\C=C(/O[Tb](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/3C11H20O2.Tb/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b3*8-7-;

InChI key

JSAHJYGCYOFNGW-LWTKGLMZSA-K

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingru Wang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 235, 118260-118260 (2020-03-29)
Eu/Tb co-doped films with low concentration gold nanorods have been prepared using the solution process. The luminescence spectra investigations indicate that the introduction of nanorods can effectively enhance the energy transfer from Tb to Eu under excitation of 292 nm, because

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service