Skip to Content
Merck
All Photos(1)

Documents

429740

Sigma-Aldrich

Cobalt(II) iodide

anhydrous, beads, −10 mesh, 99.999%

Synonym(s):

Cobalt diiodide, Cobalt iodide, Cobaltous iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoI2
CAS Number:
Molecular Weight:
312.74
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.999%

form

beads

particle size

−10 mesh

mp

>515 °C (lit.)

density

5.45 g/mL at 25 °C (lit.)

SMILES string

I[Co]I

InChI

1S/Co.2HI/h;2*1H/q+2;;/p-2

InChI key

AVWLPUQJODERGA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service