Skip to Content
Merck
All Photos(1)

Key Documents

429600

Sigma-Aldrich

Benzaldehyde-2,4-dinitrophenylhydrazone

environmental standard, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
286.24
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

environmental standard

Assay

99%

mp

239-241 °C (lit.)

application(s)

environmental

SMILES string

[O-][N+](=O)c1ccc(N\N=C\c2ccccc2)c(c1)[N+]([O-])=O

InChI

1S/C13H10N4O4/c18-16(19)11-6-7-12(13(8-11)17(20)21)15-14-9-10-4-2-1-3-5-10/h1-9,15H/b14-9+

InChI key

DZPRPFUXOZTWAJ-NTEUORMPSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service