Skip to Content
Merck
All Photos(1)

Documents

428337

Sigma-Aldrich

Cyclopentylmagnesium bromide solution

2.0 M in diethyl ether

Synonym(s):

Bromo(cyclopentyl)magnesium, Bromocyclopentylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9MgBr
CAS Number:
Molecular Weight:
173.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

bp

35 °C

density

0.996 g/mL at 25 °C

SMILES string

Br[Mg]C1CCCC1

InChI

1S/C5H9.BrH.Mg/c1-2-4-5-3-1;;/h1H,2-5H2;1H;/q;;+1/p-1

InChI key

PZOJUJCALDFFHC-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

-36.4 °F - closed cup

Flash Point(C)

-38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leandro Val Sayson et al.
Psychopharmacology, 236(7), 2201-2210 (2019-03-21)
Depressive syndrome or depression is a debilitating brain disorder affecting numerous people worldwide. Although readily available, current antidepressants have low remission rates and late onset times. Recently, N-methyl-D-aspartate (NMDA) receptor antagonists, like ketamine and methoxetamine (MXE), were found to elicit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service