427098
Terbium(III) iodide
anhydrous, flakes, 99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
TbI3
CAS Number:
Molecular Weight:
539.64
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade
anhydrous
Assay
99.99% trace metals basis
form
flakes
reaction suitability
reagent type: catalyst
core: terbium
SMILES string
I[Tb](I)I
InChI
1S/3HI.Tb/h3*1H;/q;;;+3/p-3
InChI key
OJXRJPFRTRETRN-UHFFFAOYSA-K
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service