Skip to Content
Merck
All Photos(1)

Key Documents

42322

Sigma-Aldrich

3-(Bromomethyl)benzoic acid N-succinimidylester

≥95.0%

Synonym(s):

N-Succinimidyl 3-(bromomethyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10BrNO4
CAS Number:
Molecular Weight:
312.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0%

reaction suitability

reagent type: cross-linking reagent

storage temp.

−20°C

SMILES string

BrCc1cccc(c1)C(=O)ON2C(=O)CCC2=O

InChI

1S/C12H10BrNO4/c13-7-8-2-1-3-9(6-8)12(17)18-14-10(15)4-5-11(14)16/h1-3,6H,4-5,7H2

InChI key

BHXBPWLMEXGVCV-UHFFFAOYSA-N

General description

3-(Bromomethyl)benzoic acid N-succinimidylester is an ester derivative of benzoic acid. It is an organic building block used in chemical synthesis.

Application

3-(Bromomethyl)benzoic acid N-succinimidyl ester may be used as hetero-bifunctional linker for the quaternisation of 5-(4-methoxyphenyl)2(4-pyridyl)oxazole and 2-(6-chromanyl)5(4-pyridyl)oxazole. These compounds are useful in the preparation of fluorescent stains.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of reactive fluorescent stains derived from 5(2)-aryl-2(5)-(4-pyridyl)oxazoles and bifunctionally reactive linkers.
Litak PT and Kauffman JM.
Journal of Heterocyclic Chemistry, 31(2), 457-459 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service