Skip to Content
Merck
All Photos(1)

Key Documents

421065

Sigma-Aldrich

TYZOR® GBA organic titanate

Synonym(s):

Diisopropoxytitanium bis(acetylacetonate) solution, Ti(acac)2OiPr2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]2Ti(C5H7O2)2
CAS Number:
Molecular Weight:
364.26
MDL number:
UNSPSC Code:
12352300

composition

Ti, 10%

SMILES string

CC(C)O[Ti](OC(C)C)(O\C(C)=C\C(C)=O)O\C(C)=C\C(C)=O

InChI

1S/2C5H8O2.2C3H7O.Ti/c2*1-4(6)3-5(2)7;2*1-3(2)4;/h2*3,6H,1-2H3;2*3H,1-2H3;/q;;2*-1;+4/p-2/b2*4-3+;;;

InChI key

OVSGBKZKXUMMHS-VVDZMTNVSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Lyson-Sypien et al.
Beilstein journal of nanotechnology, 8, 108-122 (2017-02-02)
The aim of this research is to study the role of nanocrystalline TiO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service