Skip to Content
Merck
All Photos(1)

Key Documents

418943

Sigma-Aldrich

2,2′-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2NHC6H3(NO2)N(CH2CH2OH)2
CAS Number:
Molecular Weight:
285.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

108-111 °C (lit.)

solubility

water: soluble(lit.)

SMILES string

OCCNc1ccc(cc1[N+]([O-])=O)N(CCO)CCO

InChI

1S/C12H19N3O5/c16-6-3-13-11-2-1-10(9-12(11)15(19)20)14(4-7-17)5-8-18/h1-2,9,13,16-18H,3-8H2

InChI key

MIWUTEVJIISHCP-UHFFFAOYSA-N

General description

2,2′-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol (HC Blue No.2) is a water-soluble purple dye.[1]

Application

2,2′-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol may be employed to improve the visibility of self-healing ureidopyrimidinone (UPy) hydrogel (poly(ethylene glycol) containing bifunctional UDP groups).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F W Kari et al.
Cell biology and toxicology, 6(2), 139-155 (1990-04-01)
Previous studies indicated that HC Blue 1 induced heptocellular carcinomas in B6C3F1 mice whereas the structurally similar nitroaromatic amine HC Blue 2 did not. In an attempt to elucidate the biochemical mechanisms responsible for their different carcinogenic potencies, comparative metabolism
HC Blue No. 2.
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 143-151 (1993-01-01)
Species comparisons regarding comparative metabolism of two structurally similar phenylenediamines (HC Blue 1 and HC Blue 2).
F Kari et al.
Progress in clinical and biological research, 340D, 305-314 (1990-01-01)
Self-Healing Supramolecular Polymers In Action.
van Gemert GML, et al.
Macromolecular Chemistry and Physics, 213(2), 234-242 (2012)
National Toxicology Program technical report series, 293, 1-192 (1985-08-01)
Toxicology and carcinogenesis studies of HC Blue No. 2 (approximately 98% pure), a semipermanent hair dye, were conducted by administering the test chemical in feed for 103 weeks to groups of 50 F344/N rats of each sex and for 104

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service