Skip to Content
Merck
All Photos(2)

Key Documents

417203

Sigma-Aldrich

R-Alpine-Borane®

97%

Synonym(s):

B-Isopinocampheyl-9-borabicyclo[3.3.1]nonane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31B
CAS Number:
Molecular Weight:
258.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

97%

optical activity

[α]21/D -22°, c = 12 in THF

bp

>55 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1[C@@H](CC2CC1C2(C)C)B3C4CCCC3CCC4

InChI

1S/C18H31B/c1-12-16-10-13(18(16,2)3)11-17(12)19-14-6-4-7-15(19)9-5-8-14/h12-17H,4-11H2,1-3H3/t12-,13+,14-,15+,16-,17-/m1/s1

InChI key

VCDGSBJCRYTLNU-PHPOFCCKSA-N

Related Categories

General description

R-Alpine-Borane® is a chiral reducing agent, synthesized from (+)-α-pinene via hydroboration.[1][2]

Application

R-Alpine-Borane® may be used in the preparation of (22R)-hydroxy-23-acetylenic steroids with high stereoselectivity.[3]
Reagent for the asymmetric reduction of a variety of prochiral ketones.[4]

Legal Information

Alpine-Borane is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Pyr. Liq. 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereocontrolled synthesis of 22-hydroxy-23-acetylenic steroids, key intermediates in steroid side chain construction. Observation of a directive effect by an a-chiral site during asymmetric reduction with-B-3-pinanyl-9-BBN (Alpine-Borane).
Midland MM and Kwon YC.
Tetrahedron Letters, 25(52), 5981-5984 (1984)
Ramachandran, P.V. et al.
Tetrahedron Asymmetry, 4, 2399-2399 (1993)
Diisopinocampheylchloroborane, a remarkably efficient chiral reducing agent for aromatic prochiral ketones.
Chandrasekharan J, et al.
The Journal of Organic Chemistry, 50(25), 5446-5448 (1985)
Matteson DS
Stereodirected Synthesis with Organoboranes, 346-347 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service