Skip to Content
Merck
All Photos(1)

Key Documents

413011

Sigma-Aldrich

Yttrium(III) iodide

anhydrous, flakes, 99.9% trace metals basis

Synonym(s):

Yttrium triiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YI3
CAS Number:
Molecular Weight:
469.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
flakes

grade

anhydrous

Assay

99.9% trace metals basis

form

flakes

reaction suitability

reagent type: catalyst
core: yttrium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

I[Y](I)I

InChI

1S/3HI.Y/h3*1H;/q;;;+3/p-3

InChI key

LFWQXIMAKJCMJL-UHFFFAOYSA-K

Application

Starting material for YBCO superconducting materials in a lower temperature preparation.[1] Applied in the solid-state synthesis of a new compound, Y21I18C14B7, which is the first example of a rare earth metal halide with interstitial halogen atoms with misfit characteristics.[2]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oeckler, O. et al.
Journal of the American Chemical Society, 38, 1767-1767 (1998)
Baney, R.H. et al.
Chemistry of Materials, 4, 984-984 (1992)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service