Skip to Content
Merck
All Photos(1)

Key Documents

410950

Sigma-Aldrich

Trimethyl-1,6-diisocyanatohexane, mixture of 2,2,4- and 2,4,4-isomers

99%

Synonym(s):

1,6-Diisocyanato-2,2,4(or 2,4,4)-trimethylhexane, 2,2,4(2,4,4)-Trimethylhexyl-1,6-diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H18N2O2
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.462 (lit.)

bp

149 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

CC(CCN=C=O)CC(C)(C)CN=C=O.CC(CN=C=O)CC(C)(C)CCN=C=O

InChI

1S/2C11H18N2O2/c1-10(7-13-9-15)6-11(2,3)4-5-12-8-14;1-10(4-5-12-8-14)6-11(2,3)7-13-9-15/h2*10H,4-7H2,1-3H3

InChI key

XLRHAAFCQUDHHF-UHFFFAOYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>235.4 °F

Flash Point(C)

> 113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service