SMILES string
O=C(\C=C(\O[Fe](O\C(=C\C(=O)c1ccccc1)c2ccccc2)O\C(=C\C(=O)c3ccccc3)c4ccccc4)c5ccccc5)c6ccccc6
Application
Air-stable, non-hygroscopic, THF-soluble catalyst for the coupling of alkenyl halides with Grignard reagents, and for the samarium(II) iodide promoted intramolecular Barbier-type reaction for the synthesis of bicyclo[m.n.l]alkan-1-ols.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of Organic Chemistry, 56, 4112-4112 (1991)
The Journal of Organic Chemistry, 40, 599-599 (1975)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service